Orders LinkContactUsLinkComponent
Spatulas, Chattaway type
Spatulas, Chattaway type
Spatula, Chattaway type, medium, 200 mm
Catalog # MURRL130/02
Supplier:   MURRAY S
Spatulas, Chattaway type
Catalog # MURRL130/02
Supplier:  MURRAY S
Spatulas, Chattaway type
Spatula, Chattaway type, medium, 200 mm
Price
403,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
Chattaway spatulas made of light or medium stainless steel.
Specifications
  • Popis:
    Spatula, Chattaway type, medium
  • Délka:
    200 mm