Syringes
Syringe, glass, center nozzle, luer slip, grad. 0,1 ml, 1 ml
Catalog # 613-3000
Supplier: MURRAY S
Jump to:
Product Details & Documents
Glass syringes, without needles.
- Centre nozzle with Luer slip fitting
Specifications
- Kapacita:1 ml
- Popis:Syringe, glass, center nozzle, luer slip, grad. 0,1 ml