Orders LinkContactUsLinkComponent
Dressing jars
Dressing jars
17, Lids, stainless steel, circular with plain rim, for jars of O.Ø 200 mm
Catalog # MURRG502/04
Supplier:   MURRAY S
Dressing jars
Catalog # MURRG502/04
Supplier:  MURRAY S
Dressing jars
17, Lids, stainless steel, circular with plain rim, for jars of O.Ø 200 mm
Price
1 140,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
Dressing jars, made of moulded glass, round.
  • Good chemical resistance
  • Lids must be ordered separately
Specifications
  • Popis:
    Lids, stainless steel, circular with plain rim, for jars of O.Ø 200 mm
  • Sekvence:
    17