Orders LinkContactUsLinkComponent
Dressing jars
Dressing jars
02, 125 mm, 125 mm
Catalog # MURRG500/04
Supplier:   MURRAY S
Dressing jars
Catalog # MURRG500/04
Supplier:  MURRAY S
Dressing jars
02, 125 mm, 125 mm
Price
1 090,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
Dressing jars, made of moulded glass, round.
  • Good chemical resistance
  • Lids must be ordered separately
Specifications
  • Ø vněj.:
    125 mm
  • Výška:
    125 mm
  • Sekvence:
    02