Funnels
01, 100 m mm
Katalogové číslo NICKFUNSS100H
Dodavatel: NICKEL ELECTRO
Přejít na:
Product Details & Documents
Stainless steel.
- Good chemical resistance, extremely high thermal resistance
- With air duct for discharging container air
- Autoclavable
Smooth internal walls. Robust and strong. Multipurpose funnel for a wide variety of uses. Suspension eye for easy storage.
Specifikace
- Vněj. Ø nálevky:100 m mm
- Sekvence:01