Orders LinkContactUsLinkComponent
Dissection sets
Dissection sets
Dissection kit, Advanced Set in PVC roll, 13 pcs.
Katalogové číslo 631-1093
Dodavatel:   MURRAY S
Dissection sets
Katalogové číslo 631-1093
Dodavatel:  MURRAY S
Dissection sets
Dissection sets
Dissection kit, Advanced Set in PVC roll, 13 pcs.
Přejít na:
Price
1 630,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
These high quality dissecting instruments are designed for everyday use. They are subject to stringent manufacturing quality procedures, and therefore suitable for surgical applications.
The Basic Set includes:
Two scalpels
Two needles
One seeker
One blunt forceps
One sharp forceps
One fine scissors
One blunt scissors
The Advanced Set includes:
Three scalpels
Two needles
One seeker
One section lifter
One blunt forceps
One sharp forceps
One sharp curved forceps
One fine scissors
One blunt scissors
One section razor
Specifikace
  • Popis:
    Dissection kit, Advanced Set in PVC roll, 13 pcs.