Orders LinkContactUsLinkComponent
Dissecting scissors, curved tip
Dissecting scissors, curved tip
Curved, sharp, fine point, 105 mm
Katalogové číslo 233-0159
Dodavatel:   MURRAY S
Dissecting scissors, curved tip
Katalogové číslo 233-0159
Dodavatel:  MURRAY S
Dissecting scissors, curved tip
Dissecting scissors, curved tip
Curved, sharp, fine point, 105 mm
Přejít na:
Price
136,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
Stainless steel.
  • Closed or open shanks
  • Curved, sharp/sharp
Specifikace
  • Type:
    Curved, sharp, fine point
  • Délka:
    105 mm