Orders LinkContactUsLinkComponent
Electrode Adapter Cables
Electrode Adapter Cables
11, Adapter, 2 mm pin tip meter input + screw cap electrode connector; detachable cable to 2,5 mm pin tip connector
Catalog # ORIO91USRF
Supplier:   Thermo Orion
Electrode Adapter Cables
Catalog # ORIO91USRF
Supplier:  Thermo Orion
Electrode Adapter Cables
11, Adapter, 2 mm pin tip meter input + screw cap electrode connector; detachable cable to 2,5 mm pin tip connector
Price
6 990,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
These cables can be used to convert an electrode with one connector to the required connector of another meter.
For use with meters that have BNC, US standard, certain LEMO, British, E DIN, Radiometer, 2 mm reference pins or just a stripped lead inputs.
Electrodes must have a US standard, screw cap, Karl Fisher, or BNC connectors.
Specifications
  • Popis:
    Adapter, 2 mm pin tip meter input + screw cap electrode connector; detachable cable to 2,5 mm pin tip connector
  • Sekvence:
    11