Orders LinkContactUsLinkComponent
Square HDPE Bottle
Square HDPE Bottle
01, 250 ml, Square bottle, HDPE
Catalog # SAMP8165A-250
Supplier:   Sampling Systems
Square HDPE Bottle
Catalog # SAMP8165A-250
Supplier:  Sampling Systems
Square HDPE Bottle
Square HDPE Bottle
01, 250 ml, Square bottle, HDPE
Price
192,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
Square HDPE bottles for use with a range of liquids and powders.
  • Different sizes available
  • Ribbed sides for easy handling
  • Available from stock
  • Fast delivery
This range of HDPE containers are perfect for use with a range of powders and liquids. They have a handy square space-saving shape and incorporate side ridges for easy handling when wet.
Specifications
  • Kapacita:
    250 ml
  • Popis:
    Square bottle
  • Materiál:
    HDPE
  • Sekvence:
    01
  • Cat. No.:
    SAMP8165A-250