Orders LinkContactUsLinkComponent
Magnetic stirring bars, triangular
Magnetic stirring bars, triangular
02, 25,32, 7,39 mm
Katalogové číslo KART781
Dodavatel:   KARTELL
Magnetic stirring bars, triangular
Katalogové číslo KART781
Dodavatel:  KARTELL
Magnetic stirring bars, triangular
Magnetic stirring bars, triangular
02, 25,32, 7,39 mm
Přejít na:
Price
126,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
PTFE coated, triangular magnetic stirring bars.
  • Scraping action ideal for dissolving solids and mixing sediments
  • Angled surfaces create high turbulence even at low speeds
  • Suitable for food stuff
Specifikace
  • Ø:
    7,39 mm
  • Délka:
    25,32
  • Sekvence:
    02