Orders LinkContactUsLinkComponent
Filter discs
Filter discs
09, 1, 30 mm
Catalog # LENZ07180130
Supplier:   Lenz Laborglas
Filter discs
Catalog # LENZ07180130
Supplier:  Lenz Laborglas
Filter discs
Filter discs
09, 1, 30 mm
Price
1 340,00 Kč
Log in for availability
1 * 10 KS
Product Details & Documents
Glass filter discs centred with unfused rim.
Documents
Specifications
  • Ø:
    30 mm
  • Porozita:
    1
  • Sekvence:
    09