Orders LinkContactUsLinkComponent
Filter discs
01, 0, 10 mm
Catalog # LENZ07180010
Supplier:   Lenz Laborglas
Filter discs
Catalog # LENZ07180010
Supplier:  Lenz Laborglas
Filter discs
Filter discs
01, 0, 10 mm
Price
1 160,00 Kč
Log in for availability
1 * 10 KS
Product Details & Documents
Glass filter discs centred with unfused rim.
Specifications
  • Ø:
    10 mm
  • Porozita:
    0
  • Sekvence:
    01