Clamps
01, Flat flanges DN 100
Katalogové číslo LENZ06247010
Dodavatel: Lenz Laborglas
Přejít na:
Product Details & Documents
Stainless steel.
- Excellent chemical resistance
- Excellent temperature resistance
For fixing lids onto reaction vessels, available for flat flanges LF100-200.
Specifikace
- Určeno pro:Flat flanges DN 100
- Sekvence:01