Flat flange lids, centre neck, 1 angled side neck
laboratory flange LF 150, 2 Necks, NS 45/40 (centre), 29/32 NS, 140 mm
Catalog # LENZ06102464
Supplier: Lenz Laborglas
Jump to:
Product Details & Documents
DURAN®, Borosilicate glass.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
Flange lids for reaction vessels, with laboratory flange LF.
Documents
BSE TSE Glass Columns
Certifikáty BSE/TSE
Specifications
- Type:2 Necks
- S:laboratory flange LF 150
- Hrdlo:NS 45/40 (centre)
- Boční hrdla:29/32 NS
- Celková výška:140 mm