Orders LinkContactUsLinkComponent
Vacuum stopcocks
Vacuum stopcocks
13, Vacuum stopcocks, one way, right-angled, 6,0 mm, 21,5 NS
Katalogové číslo 189-2123
Dodavatel:   Lenz Laborglas
Vacuum stopcocks
Katalogové číslo 189-2123
Dodavatel:  Lenz Laborglas
Vacuum stopcocks
Vacuum stopcocks
13, Vacuum stopcocks, one way, right-angled, 6,0 mm, 21,5 NS
Přejít na:
Price
2 190,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
Vacuum stopcocks, DURAN® with side arms and hollow NS glass plug.
  • Excellent chemical resistance
  • Excellent temperature resistance
Leakage rate according to DIN 12540-1.
Documents
Specifikace
  • Ø otvoru:
    6,0 mm
  • Popis:
    Vacuum stopcocks, one way, right-angled
  • Sekvence:
    13
  • Kohout:
    21,5 NS