Orders LinkContactUsLinkComponent
Vacuum stopcocks
Vacuum stopcocks
09, Vacuum stopcocks, one way, straight, 8,0 mm, 24 NS
Katalogové číslo LENZ02902408
Dodavatel:   Lenz Laborglas
Vacuum stopcocks
Katalogové číslo LENZ02902408
Dodavatel:  Lenz Laborglas
Vacuum stopcocks
Vacuum stopcocks
09, Vacuum stopcocks, one way, straight, 8,0 mm, 24 NS
Přejít na:
Price
2 770,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
Vacuum stopcocks, DURAN® with side arms and hollow NS glass plug.
  • Excellent chemical resistance
  • Excellent temperature resistance
Leakage rate according to DIN 12540-1.
Documents
Specifikace
  • Ø otvoru:
    8,0 mm
  • Popis:
    Vacuum stopcocks, one way, straight
  • Sekvence:
    09
  • Kohout:
    24 NS