Automatic burettes, Schilling pattern, Class B
50 ml, 0,1 ml
Katalogové číslo KART1981
Dodavatel: KARTELL
Přejít na:
Product Details & Documents
PMP/PE, class B.
- Push button for drop-by- drop delivery
- PE reservoir with stable LDPE base
According to DIN 12.700 standards, class B
Certifications: According to DIN 12.700 standards, class B
Specifikace
- Kapacita:50 ml
- Dělení:0,1 ml