Orders LinkContactUsLinkComponent
Spray bottles
Spray bottles
03, 1000 ml, Spray bottle, Bezbarvá
Katalogové číslo VITL95286
Dodavatel:   VITLAB
Spray bottles
Katalogové číslo VITL95286
Dodavatel:  VITLAB
Spray bottles
Spray bottles
03, 1000 ml, Spray bottle, Bezbarvá
Přejít na:
Price
305,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
White or translucent made from PP.
  • Excellent chemical resistance
  • Excellent temperature resistance
Sprayer insert with stable, smoothly operated pump trigger and adjustable spray nozzle, which can be regulated from the finest mist (nebulising) to a precise liquid jet. Range: approx. 3 to 4 metres. Ideal for spraying detergents or disinfectants, especially into difficultly accessible areas, as well as applications in thin layer chromatography.
Specifikace
  • Kapacita:
    1000 ml
  • Barva:
    Bezbarvá
  • Popis:
    Spray bottle
  • Sekvence:
    03