2-Bromophenol ≥98%
3401, Glass bottle for solids, 100 g
Purity: 98% CAS No.: 95-56-7 Molecular Formula: C6H5BrO Formula Weight: 173.01 Flash Point: 89°(192°F) Refractive Index: 1.5890 Odour: Phenol-like Melting Point: 5° Boiling Point: 194-195° Beilstein Reference: 1905115 Merck Reference: 14,1428 UN No.: 3082 MDL No.: MFCD00002146
- :95-56-7
- :C₆H₅BrO
- :MFCD00002146
- :173.01
- :14,1428
- :194…195 °C
- :5 °C
- :1,636
- :92 °C (192 °F)
- :Ambient
- :3082
- :9,III
- :Density: 1,49 g/cm³ (20 °C)
- :202-432-7
- Typ balení:Glass bottle for solids
- Bal.:100 g
- Sekvence:3401
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.