Orders LinkContactUsLinkComponent
2-Bromophenol ≥98%
3401, Glass bottle for solids, 100 g
  A14112.22
2-Bromophenol ≥98%
  A14112.22
 :  95-56-7
2-Bromophenol ≥98%
2-Bromophenol ≥98%
3401, Glass bottle for solids, 100 g
 :  95-56-7
13 430,00 Kč
1 * 100 g
Purity: 98% CAS No.: 95-56-7 Molecular Formula: C6H5BrO Formula Weight: 173.01 Flash Point: 89°(192°F) Refractive Index: 1.5890 Odour: Phenol-like Melting Point: 5° Boiling Point: 194-195° Beilstein Reference: 1905115 Merck Reference: 14,1428 UN No.: 3082 MDL No.: MFCD00002146
  •  :95-56-7
  •  :C₆H₅BrO
  •  :MFCD00002146
  •  :173.01
  •  :14,1428
  •  :194…195 °C
  •  :5 °C
  •  :1,636
  •  :92 °C (192 °F)
  •  :Ambient
  •  :3082
  •  :9,III
  •  :Density: 1,49 g/cm³ (20 °C)
  •  :202-432-7
  • Typ balení:
    Glass bottle for solids
  • Bal.:
    100 g
  • Sekvence:
    3401