Hydrogensíran draselný ≥97%
3645, Glass bottle for solids, 500 g
Dodavatel: Thermo Fisher Scientific
Hydrogensíran draselný ≥97%
Katalogové číslo A12428.36
Dodavatel: Thermo Fisher Scientific
Číslo CAS: 7646-93-7
Přejít na:
Product Details & Documents
Purity: 97% CAS No.: 7646-93-7 Molecular Formula: HKO4S Formula Weight: 136.17 Odour: Odourless Melting Point: 195-197° dec. UN No.: 2509 MDL No.: MFCD00011404
Synonyma: Potassium bisulphate
- Číslo CAS:7646-93-7
- Vzorec:KHSO₄
- MDL číslo:MFCD00011404
- Molecular Weight:136.17
- Merck Index:14,7613
- Bod tání:195…197 °C (rozkl.)
- Hustota:2,240
- Teplota pro skladování:Ambient
- EINECS:231-594-1
- UN:2509
- ADR:8,II
- :Density: 2,32 g/cm³ (20 °C)
Documents
Specifikace
Specifikace
- Typ balení:Glass bottle for solids
- Bal.:500 g
- Sekvence:3645
Specifikace
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



