Zadání objednávky
Czech Republic
Orders LinkContactUsLinkComponent
Hydrogensíran draselný ≥97%
3900, Glass bottle for solids, 10.000 g
  A12428.0C
Hydrogensíran draselný ≥97%
  A12428.0C
 :  7646-93-7
Hydrogensíran draselný ≥97%
Hydrogensíran draselný ≥97%
3900, Glass bottle for solids, 10.000 g
 :  7646-93-7
15 080,00 Kč
1 * 10.000 g
Purity: 97% CAS No.: 7646-93-7 Molecular Formula: HKO4S Formula Weight: 136.17 Odour: Odourless Melting Point: 195-197° dec. UN No.: 2509 MDL No.: MFCD00011404
 :  Potassium bisulphate
  •  :7646-93-7
  •  :KHSO₄
  •  :MFCD00011404
  •  :136.17
  •  :14,7613
  •  :195…197 °C (rozkl.)
  •  :2,240
  •  :Ambient
  •  :231-594-1
  •  :2509
  •  :8,II
  •  :Density: 2,32 g/cm³ (20 °C)
  • Typ balení:
    Glass bottle for solids
  • Bal.:
    10.000 g
  • Sekvence:
    3900
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.