Hydrogensíran draselný ≥97%
3900, Glass bottle for solids, 10.000 g
Purity: 97% CAS No.: 7646-93-7 Molecular Formula: HKO4S Formula Weight: 136.17 Odour: Odourless Melting Point: 195-197° dec. UN No.: 2509 MDL No.: MFCD00011404
: Potassium bisulphate
- :7646-93-7
- :KHSO₄
- :MFCD00011404
- :136.17
- :14,7613
- :195…197 °C (rozkl.)
- :2,240
- :Ambient
- :231-594-1
- :2509
- :8,II
- :Density: 2,32 g/cm³ (20 °C)
- Typ balení:Glass bottle for solids
- Bal.:10.000 g
- Sekvence:3900