ACES ≥99%
3308, Glass bottle for solids, 25 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Purity: 99% CAS No.: 7365-82-4 Molecular Formula: H2NCOCH2NHCH2CH2SO3H Formula Weight: 182.20 Melting Point: ca 275° dec. Beilstein Reference: 2253770 Merck Reference: 14,36 MDL No.: MFCD00008030
Synonyms: ACES, N-(Carbamoylmethyl)taurine, N-(Carbamoylmethyl)-2-aminoethanesulfonic acid, N-(2-Acetamido)-2-aminoethanesulphonic acid, N-(Carbamoylmethyl)-2-aminoethanesulphonic Acid, 2-[(Carbamoylmethyl)amino]ethanesulfonic acid
- CAS Number:7365-82-4
- Formula:H₂NCOCH₂NHCH₂CH₂SO₃H
- MDL Number:MFCD00008030
- Molecular Weight:182.2
- Merck Index:14,36
- Melting Pt:∼ 275 °C (rozkl.)
- Storage Temperature:Ambient
- EINECS:230-908-4
Documents
Specifications
Specifications
- Typ balení:Glass bottle for solids
- Bal.:25 g
- Sekvence:3308



