ethylformiát ≥97%
2895, Glass bottle, 2.500 mL
Dodavatel: Thermo Fisher Scientific
Přejít na:
Product Details & Documents
Purity: 97% CAS No.: 109-94-4 Molecular Formula: C3H6O2 Formula Weight: 74.08 Storage Temperature: 2°C to 8°C Flash Point: -19°(-2°F) Physical Form: Liquid Density: 0.921 Refractive Index: 1.3600 Appearance: Colourless Odour: Fruit-like pH: 4.1 Melting Point: -81° Boiling Point: 52-54° Beilstein Reference: 906769 Merck Reference: 14,3807 UN No.: 1190 MDL No.: MFCD00003294
Synonyma: Formic acid ethyl ester
- Číslo CAS:109-94-4
- Vzorec:C₃H₆O₂
- MDL číslo:MFCD00003294
- Molecular Weight:74.08
- Merck Index:14,3807
- Bod varu:52…54 °C
- Bod tání:–81 °C
- Hustota:0,921
- Bod vzplanutí:–19 °C (–2 °F)
- Teplota pro skladování:Refrigerator
- UN:1190
- ADR:3,II
- :Density: 0,9208 g/cm³ (20 °C)
- EINECS:203-721-0
Documents
Specifikace
Specifikace
- Typ balení:Glass bottle
- Bal.:2.500 mL
- Sekvence:2895



