Orders LinkContactUsLinkComponent
Cables for Electrodes
Cables for Electrodes
20, Plug cable combination, K10X (coaxial electrode plug, 10 m coaxial cable, free end), 10 m
Catalog # 662-2186
Supplier:   SI Analytics
Cables for Electrodes
Catalog # 662-2186
Supplier:  SI Analytics
Cables for Electrodes
Cables for Electrodes
20, Plug cable combination, K10X (coaxial electrode plug, 10 m coaxial cable, free end), 10 m
Price
3 330,00 Kč
Log in for availability
1 * 1 KS
Specifications
  • Popis:
    Plug cable combination, K10X (coaxial electrode plug, 10 m coaxial cable, free end)
  • Délka:
    10 m
  • Sekvence:
    20