Orders LinkContactUsLinkComponent
Wall - mounted cabinets
Wall - mounted cabinets
Wall-mounted cabinet, with 6 shelves, 2, 920×300×800 mm
Katalogové číslo 132-0401
Dodavatel:   Palbam
Wall - mounted cabinets
Katalogové číslo 132-0401
Dodavatel:  Palbam
Wall - mounted cabinets
Wall - mounted cabinets
Wall-mounted cabinet, with 6 shelves, 2, 920×300×800 mm
Přejít na:
Price
82 430,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
Stainless steel 304 with electropolished finish
  • High-quality finish
Balení: Typically available from stock for direct shipment from the supplier. Shipped flat-packed for simple assembly by the customer. Assembly instructions included.
Specifikace
  • Popis:
    Wall-mounted cabinet, with 6 shelves
  • Š×H×V:
    920×300×800 mm
  • Dveře:
    2