Orders LinkContactUsLinkComponent
Wall - mounted cabinets
Wall - mounted cabinets
Wall-mounted cabinet, with 3 shelves, 1, 460×300×800 mm
Katalogové číslo 132-0400
Dodavatel:   Palbam
Wall - mounted cabinets
Katalogové číslo 132-0400
Dodavatel:  Palbam
Wall - mounted cabinets
Wall - mounted cabinets
Wall-mounted cabinet, with 3 shelves, 1, 460×300×800 mm
Přejít na:
Price
48 890,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
Stainless steel 304 with electropolished finish
  • High-quality finish
Balení: Typically available from stock for direct shipment from the supplier. Shipped flat-packed for simple assembly by the customer. Assembly instructions included.
Specifikace
  • Popis:
    Wall-mounted cabinet, with 3 shelves
  • Š×H×V:
    460×300×800 mm
  • Dveře:
    1