Orders LinkContactUsLinkComponent
Basic cleanroom benches
Basic cleanroom benches
03, Basic cleanroom bench, 1000×350×450 mm
Katalogové číslo 134-0498
Dodavatel:   Palbam
Basic cleanroom benches
Katalogové číslo 134-0498
Dodavatel:  Palbam
Basic cleanroom benches
Basic cleanroom benches
03, Basic cleanroom bench, 1000×350×450 mm
Přejít na:
Price
30 680,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
Cleanroom benches in stainless steel 304 with electropolished finish.
  • Rounded edges
  • Height adjustable legs
  • Leg covers (covers unsightly bolt connections to floor)
  • No snag points
  • Ultra-clean
Informace o dodávce: Typically available from stock for direct shipment from the supplier.
Specifikace
  • Popis:
    Basic cleanroom bench
  • Sekvence:
    03
  • Š×H×V:
    1000×350×450 mm