Orders LinkContactUsLinkComponent
Basic cleanroom benches
Basic cleanroom benches
02, Basic cleanroom bench, 800×350×450 mm
Catalog # 134-0497
Supplier:   Palbam
Basic cleanroom benches
Catalog # 134-0497
Supplier:  Palbam
Basic cleanroom benches
Basic cleanroom benches
02, Basic cleanroom bench, 800×350×450 mm
Price
26 380,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
Cleanroom benches in stainless steel 304 with electropolished finish.
  • Rounded edges
  • Height adjustable legs
  • Leg covers (covers unsightly bolt connections to floor)
  • No snag points
  • Ultra-clean
Informace o dodávce: Typically available from stock for direct shipment from the supplier.
Specifications
  • Popis:
    Basic cleanroom bench
  • Sekvence:
    02
  • Š×H×V:
    800×350×450 mm