Orders LinkContactUsLinkComponent
Hexagonal nuts with inner thread
Hexagonal nuts with inner thread
hexagonal nut with inner thread, for Ø 36 mm, ¾", PE
Katalogové číslo BURK8537-0034
Dodavatel:   BURKLE
Hexagonal nuts with inner thread
Katalogové číslo BURK8537-0034
Dodavatel:  BURKLE
Hexagonal nuts with inner thread
Hexagonal nuts with inner thread
hexagonal nut with inner thread, for Ø 36 mm, ¾", PE
Přejít na:
Price
100,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
HDPE, white.
  • Good chemical resistance
  • Good temperature resistance
Hexagonal nut with inner thread.
Specifikace
  • Popis:
    hexagonal nut with inner thread, for Ø 36 mm
  • Materiál:
    PE
  • Závit:
    ¾"