pH electrodes, InLab®, Expert Series
07, InLab® Expert Go, InLab® Expert Go, 0 - 14, 0...+100 °C, PEEK, XEROLYT® polymer, 12×120 mm, BNC/RCA (Cinch), 1,8 m cable (IP67), Portable applications in the lab and outdoors
Dodavatel: Mettler - Toledo
Přejít na:
Product Details & Documents
Ideal for critical samples such as suspensions or samples of unknown composition (e.g. wastewater).
- ARGENTHAL™ reference system with silver-ion trap
- Solid XEROLYT® polymer electrolyte: No need for a junction (no possibility of contamination)
- Unbreakable PEEK shaft
- Conform to RoHS ('Restriction of Hazardous Substances' in electrical and electronic equipment) and WEEE ('Waste Electrical and Electronic Equipment')
pH range: 0 to 14
Temperature range: 0 to 100 °C
Temperature probe: NTC30 kOhm for InLab® Expert Pro and InLab® Expert NTC30; PT1000 for InLab® Expert DIN and InLab® Expert PT1000
: Supplied with test certificate.
Documents
Certifikáty
Hledat podle čísla šaržeMETT51343101_TDS_EN
Technický list
Mettler InLab® Sensors
Produktová literatura
Specifikace
- Připojení:BNC/RCA (Cinch), 1,8 m cable (IP67)
- Ø×D:12×120 mm
- Určeno pro:Portable applications in the lab and outdoors
- Type:InLab® Expert Go
- Popis:InLab® Expert Go
- Materiál:PEEK
- Sekvence:07
- Elektrolyt:XEROLYT® polymer
- Tepl.:0...+100 °C
- pH:0 - 14



