Orders LinkContactUsLinkComponent
Temperature indicating tubes
01, For autoclaves up to 125 °C
Katalogové číslo KOEHK-1250
Temperature indicating tubes
Katalogové číslo KOEHK-1250
Temperature indicating tubes
Temperature indicating tubes
01, For autoclaves up to 125 °C
Přejít na:
Price
6 180,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
Temperature indicating tubes are used for autoclaves and hot air sterilisers.
Sterilisation is detected by the colour change of the control tube from red to green. The tubes should be placed as close as possible next to or in the load.
Specifikace
  • Popis:
    For autoclaves up to 125 °C
  • Sekvence:
    01