Orders LinkContactUsLinkComponent
Temperature indicating tubes
Temperature indicating tubes
03, For hot-air sterilization 125 °C/15 min
Katalogové číslo 391-0084
Temperature indicating tubes
Katalogové číslo 391-0084
Temperature indicating tubes
Temperature indicating tubes
03, For hot-air sterilization 125 °C/15 min
Přejít na:
Price
6 370,00 Kč
Přihlásit se pro dostupnost
1 * 100 KS
Product Details & Documents
Temperature indicating tubes are used for autoclaves and hot air sterilisers.
Sterilisation is detected by the colour change of the control tube from red to green. The tubes should be placed as close as possible next to or in the load.
Specifikace
  • Popis:
    For hot-air sterilization 125 °C/15 min
  • Sekvence:
    03