Orders LinkContactUsLinkComponent
Dissecting forceps, simple type
Dissecting forceps, simple type
02, Titanium, curved, with fine sharp tips, 110 mm
Catalog # HAMMHTC091-10
Supplier:   HAMMACHER KARL
Dissecting forceps, simple type
Catalog # HAMMHTC091-10
Supplier:  HAMMACHER KARL
Dissecting forceps, simple type
Dissecting forceps, simple type
02, Titanium, curved, with fine sharp tips, 110 mm
Price
1 140,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
Simple dissecting forceps made from titanium.
Specifications
  • Type:
    Titanium, curved, with fine sharp tips
  • Délka:
    110 mm
  • Sekvence:
    02