Nalgene™ Screw Caps
21, Linerless, 13-415, Modrá, PP
Katalogové číslo 215-1610
Dodavatel: Thermo Fisher Scientific
Nalgene™ Screw Caps
Katalogové číslo 215-1610
Dodavatel: Thermo Fisher Scientific
Přejít na:
Product Details & Documents
PP, coloured.
- Very good chemical resistance
- Heat resistant up to 135 °C
- Autoclavable
Screw caps without inserts for Nalgene® narrow and wide mouth bottles, as well as Erlenmeyer flasks with thread.
Specifikace
- Velikost víčka:13-415
- Barva:Modrá
- Popis:Linerless
- Materiál:PP
- Sekvence:21
- Cat. No.:215-1610
- Certifikát ochrany životního prostředí:NAR Certificate of Retirement
- Environmentally Preferable:Low Manufacturing Impact