Nalgene™ Screw Caps
18, Linerless, 24-415, Červený, PP
215-1607
PP, coloured.
- Very good chemical resistance
- Heat resistant up to 135 °C
- Autoclavable
Screw caps without inserts for Nalgene® narrow and wide mouth bottles, as well as Erlenmeyer flasks with thread.
- Velikost víčka:24-415
- Barva:Červený
- Popis:Linerless
- Materiál:PP
- Sekvence:18
- Cat. No.:215-1607
- Certifikát ochrany životního prostředí:NAR Certificate of Retirement
- Environmentally Preferable:Low Manufacturing Impact