Nalgene™ Screw Caps
06, Linerless, 13-415, Bílá, PP
Katalogové číslo 215-1599
Dodavatel: Thermo Fisher Scientific
Nalgene™ Screw Caps
Katalogové číslo 215-1599
Dodavatel: Thermo Fisher Scientific
Přejít na:
Product Details & Documents
PP, coloured.
- Very good chemical resistance
- Heat resistant up to 135 °C
- Autoclavable
Screw caps without inserts for Nalgene® narrow and wide mouth bottles, as well as Erlenmeyer flasks with thread.
Specifikace
- Velikost víčka:13-415
- Barva:Bílá
- Popis:Linerless
- Materiál:PP
- Sekvence:06
- Cat. No.:215-1599
- Certifikát ochrany životního prostředí:NAR Certificate of Retirement
- Environmentally Preferable:Low Manufacturing Impact