Nalgene™ Screw Caps
01, Linerless, 13-415, Přírodní, PP
Catalog # 215-1591
Supplier: Thermo Fisher Scientific
Nalgene™ Screw Caps
Catalog # 215-1591
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
PP, coloured.
- Very good chemical resistance
- Heat resistant up to 135 °C
- Autoclavable
Screw caps without inserts for Nalgene® narrow and wide mouth bottles, as well as Erlenmeyer flasks with thread.
Specifications
- Velikost víčka:13-415
- Barva:Přírodní
- Popis:Linerless
- Materiál:PP
- Sekvence:01
- Cat. No.:215-1591
- Certifikát ochrany životního prostředí:NAR Certificate of Retirement
- Environmentally Preferable:Low Manufacturing Impact