Orders LinkContactUsLinkComponent
Transport containers
Transport containers
02, Slide mailers, PP, 87×47×16 mm, 2
Catalog # DELT900025
Supplier:   DELTALAB
Transport containers
Catalog # DELT900025
Supplier:  DELTALAB
Transport containers
Transport containers
02, Slide mailers, PP, 87×47×16 mm, 2
Price
1 770,00 Kč
Log in for availability
1 * 100 KS
Product Details & Documents
Designed for slide transport and protection.
  • Made of polypropylene
  • Easy snap-lock
  • Not indicated for liquid samples
Compartment shape ensures tight fit: no risk of damaging slides and to send by mail, recommend to use special envelopes.
Documents
Specifications
  • Počet sklíček:
    2
  • Popis:
    Slide mailers
  • Materiál:
    PP
  • Sekvence:
    02
  • Š×H×V:
    87×47×16 mm