Orders LinkContactUsLinkComponent
Transport containers
Transport containers
03, Slide mailers, PP, 84×71×6 mm, 2
Katalogové číslo DELT979930
Dodavatel:   DELTALAB
Transport containers
Katalogové číslo DELT979930
Dodavatel:  DELTALAB
Transport containers
Transport containers
03, Slide mailers, PP, 84×71×6 mm, 2
Přejít na:
Price
8 700,00 Kč
Přihlásit se pro dostupnost
1 * 250 KS
Product Details & Documents
Designed for slide transport and protection.
  • Made of polypropylene
  • Easy snap-lock
  • Not indicated for liquid samples
Compartment shape ensures tight fit: no risk of damaging slides and to send by mail, recommend to use special envelopes.
Documents
Specifikace
  • Počet sklíček:
    2
  • Popis:
    Slide mailers
  • Materiál:
    PP
  • Sekvence:
    03
  • Š×H×V:
    84×71×6 mm