Orders LinkContactUsLinkComponent
Trypsinising flasks
Trypsinising flasks
01, 250 ml
Catalog # WITG5.507.303
Supplier:   witeg Labortechnik
Trypsinising flasks
Catalog # WITG5.507.303
Supplier:  witeg Labortechnik
Trypsinising flasks
Trypsinising flasks
01, 250 ml
Price
1 330,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
Trypsinising flasks, graduated, designed for converting tissue fragments and homogenates into cell suspensions by trypsin digestion of connective proteins. Flutes in flask enhance vigorous agitation.
Specifications
  • Kapacita:
    250 ml
  • Sekvence:
    01