[EN]BUFFER SOLUTION PH 9.00 1 * 900 mL
Katalogové číslo METR6.2307.220
Dodavatel: METROHM
Frequently Bought Together
pH buffer solutions
Dodavatel: Mettler - Toledo
Buffer solutions, Certipur®, colour coded, Supelco®
Dodavatel: Merck
Roztoky pufrů v sáčcích, Certipur®, Supelco®
Dodavatel: Merck
pH buffers, ready to use
Dodavatel: WTW
VWR® AVS TITRINORM, pufrovací roztoky, 20 °C
Dodavatel: VWR Chemicals
VWR®, Primary colour solutions, Reagent Ph. Eur.
Dodavatel: VWR Chemicals
Buffer solutions in sachets
Dodavatel: Mettler - Toledo
[EN]BUFFER SOLUTION PH 7.00 1 * 900 mL
Dodavatel: METROHM
pH buffer, single-use, SINGLET™
Dodavatel: Hach
Benzofenon, Supelco®
Dodavatel: Merck