Glove liners, polyester, BCR™
02, Half-finger liners, L, Bílá, 10 pairs/pack
Katalogové číslo 112-0616
Dodavatel: BERKSHIRE
Přejít na:
Product Details & Documents
Continuous-filament knitted, these ambidextrous polyester gloves provide protection and comfort during extended wear. Ideal under latex, PVC, or other outer gloves to absorb perspiration from hands during periods of extended wear.
- Seamless construction prevents unravelling and release of unwanted particles into critical environments
- Launderable for repeated use
- Comfortable half-finger design provides greater dexterity than a full-finger glove liner and has reduced courses in the finger and cuff
- Full-finger style provides complete protection from irritation caused by latex and PVC gloves
Specifikace
- Velikost rukavic:L
- Balené:10 pairs/pack
- Barva:Bílá
- Sekvence:02
- Model:Half-finger liners
- Cat. No.:112-0616
- Obouruké/jednoruké:Obouruké
- Jednorázové/znovupoužitelné:opakovatelně použitelné