Orders LinkContactUsLinkComponent
Latex tests
Latex tests
23, Salmonella O Polyvalent (Group A-S), 2 mL
Catalog # OXOIR30858201
Latex tests
Catalog # OXOIR30858201
CAS Number:  
Latex tests
23, Salmonella O Polyvalent (Group A-S), 2 mL
CAS Number:  
Price
3 680,00 Kč
Log in for availability
1 * 2 mL
Product Details & Documents
Documents
Specifications
Specifications
  • Bal.:
    2 mL
  • Popis:
    Salmonella O Polyvalent (Group A-S)
  • Sekvence:
    23