Orders LinkContactUsLinkComponent
Solid phase microextraction fibers
Solid phase microextraction fibers
24, Kit 2: carboxen/PDMS, PDMS/DVB, and polyacrylate, for volatiles or polar organics, for autosampler use
Katalogové číslo VARISU57321U
Dodavatel:   VARIAN
Solid phase microextraction fibers
Katalogové číslo VARISU57321U
Dodavatel:  VARIAN
Solid phase microextraction fibers
24, Kit 2: carboxen/PDMS, PDMS/DVB, and polyacrylate, for volatiles or polar organics, for autosampler use
Price
Poptat cenu
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
Solid phase microextraction (SPME) is a technique for extracting analytes from solid, liquid or gaseous samples by adsorbing them onto the SPME fiber and then desorbing them into an inlet, either on a gas chromatograph (GC) or an HPLC system. SPME is amenable to automation using an autosampler or it can be performed manually as well.
Solid phase microextraction kits: Kits are also available to support method development, offering a variety of fiber types and configurations within a single kit. SPME fiber kits contain three fibers.
Specifikace
  • Popis:
    Kit 2: carboxen/PDMS, PDMS/DVB, and polyacrylate, for volatiles or polar organics, for autosampler use
  • Sekvence:
    24
Accessories
Informace o produktu
  • Katalogové číslo
    Sekvence
    Dostupnost
    Price
  • Merlin microseal, SPME replacement seal, 23 gauge
    Merlin microseal, SPME replacement seal, 23 gauge
    VARI392609902
    32
    Přihlásit se pro dostupnost
    1 * 1 KS - 13 268,31 Kč
Informace o produktu
  • Katalogové číslo
    Sekvence
    Dostupnost
    Price
  • SPME fiber holder for CTC autosampler
    SPME fiber holder for CTC autosampler
    VARISU57347U
    34
    Přihlásit se pro dostupnost
    1 * 1 KS
Informace o produktu
  • Katalogové číslo
    Sekvence
    Dostupnost
    Price
  • SPME stand, 15 ml
    SPME stand, 15 ml
    VARISU57357U
    33
    Přihlásit se pro dostupnost
    1 * 1 KS
Informace o produktu
  • Katalogové číslo
    Sekvence
    Dostupnost
    Price
  • SPME inlet guide for manual injection
    SPME inlet guide for manual injection
    VARISU57356U
    36