Orders LinkContactUsLinkComponent
Solid phase microextraction fibers
Solid phase microextraction fibers
29, Kit 2: carboxen/PDMS, PDMS/DVB, and polyacrylate, for volatiles or polar organics, for manual use
Catalog # VARISU57320U
Supplier:   VARIAN
Solid phase microextraction fibers
Catalog # VARISU57320U
Supplier:  VARIAN
Solid phase microextraction fibers
29, Kit 2: carboxen/PDMS, PDMS/DVB, and polyacrylate, for volatiles or polar organics, for manual use
Price
Inquire For Price
Log in for availability
1 * 1 KS
Product Details & Documents
Solid phase microextraction (SPME) is a technique for extracting analytes from solid, liquid or gaseous samples by adsorbing them onto the SPME fiber and then desorbing them into an inlet, either on a gas chromatograph (GC) or an HPLC system. SPME is amenable to automation using an autosampler or it can be performed manually as well.
Solid phase microextraction kits: Kits are also available to support method development, offering a variety of fiber types and configurations within a single kit. SPME fiber kits contain three fibers.
Specifications
  • Popis:
    Kit 2: carboxen/PDMS, PDMS/DVB, and polyacrylate, for volatiles or polar organics, for manual use
  • Sekvence:
    29
Accessories
Product Information
  • Catalog #
    Sekvence
    Popis
    Availability
    Price
  • VARI392609902
    32
    Merlin microseal, SPME replacement seal, 23 gauge
    Log in for availability
    1 * 1 KS - 13 268,31 Kč
Product Information
  • Catalog #
    Sekvence
    Popis
    Availability
    Price
  • VARISU57347U
    34
    SPME fiber holder for CTC autosampler
    Log in for availability
    1 * 1 KS
Product Information
  • Catalog #
    Sekvence
    Popis
    Availability
    Price
  • VARISU57357U
    33
    SPME stand, 15 ml
    Log in for availability
    1 * 1 KS
Product Information
  • Catalog #
    Sekvence
    Popis
    Availability
    Price
  • VARISU57356U
    36
    SPME inlet guide for manual injection