Orders LinkContactUsLinkComponent
Solid phase extraction cartridges, Bond Elut™ Florisil
Solid phase extraction cartridges, Bond Elut™ Florisil
15, Bond Elut™ Jr, Florisil, 200 µm, 1000 mg
Catalog # VARI12166014B
Supplier:   VARIAN
Solid phase extraction cartridges, Bond Elut™ Florisil
Catalog # VARI12166014B
Supplier:  VARIAN
Solid phase extraction cartridges, Bond Elut™ Florisil
15, Bond Elut™ Jr, Florisil, 200 µm, 1000 mg
Price
12 368,07 Kč
Log in for availability
1 * 100 KS
Product Details & Documents
Florisil is a magnesia-loaded silica gel, extremely polar in nature and ideal for the isolation of polar compounds from non-polar matrices.
  • Pesticide residue (PR) grade
  • Fast flow, ideal for viscous samples
The larger particle size of the sorbent enables flow for large sample volumes and is therefore an attractive alternative to silica if the sample matrix is particularly viscous.
Specifications
  • Velikost částic:
    200 µm
  • Popis:
    Bond Elut™ Jr
  • Sekvence:
    15
  • Výška vrstvy:
    1000 mg
  • Fáze:
    Florisil