Orders LinkContactUsLinkComponent
Lids for buckets
Lids for buckets
06, Lever lid for 10 L bucket, PP, water proof
Catalog # RIXI8-7100-100
Lids for buckets
Catalog # RIXI8-7100-100
Lids for buckets
Lids for buckets
06, Lever lid for 10 L bucket, PP, water proof
Price
2 640,00 Kč
Log in for availability
1 * 48 KS
Product Details & Documents
Water proof lids for buckets.
Ordering information: Buckets should be ordered separately.
Specifications
  • Popis:
    Lever lid for 10 L bucket, PP, water proof
  • Sekvence:
    06