Pierce™ BCA Protein Assay Kits - Reducing Agent Compatible
01, BCA protein assay kit - reducing agent compatible, 1 KIT
Katalogové číslo PIER23250
Dodavatel: Thermo Fisher Scientific
Pierce™ BCA Protein Assay Kits - Reducing Agent Compatible
Katalogové číslo PIER23250
Dodavatel: Thermo Fisher Scientific
Číslo CAS:
Přejít na:
Product Details & Documents
This BCA Protein Assay Kit - Reducing Agent Compatible enables the measurement of protein concentration in samples that contain thiol-reductants dithiothreitol (DTT) and 2-mercaptoethanol (BME).
- Compatible - assay samples that contain up to 5 mM DTT, 35 mM BME, or 10 mM TCEP
- BCA technology - only a slight modification of the standard BCA Protein Assay protocol (15 minute incubation with Compatibility Reagent); no precipitation steps required
- Small samples - requires only 25 µl (standard kit) or less than 10 µl (microplate kit) of sample
- Colorimetric - measure with a standard spectrophotometer or plate reader (562 nm)
- Excellent uniformity - exhibits less protein-to-protein variation than dye-binding methods
- High linearity - linear working range for BSA equals 125 to 2000 µg/ml
: BCA Protein Assay Kit - RAC contains sufficient reagents for 250 tube assays with 25 µl samples; BCA Reagent A, 250 ml; BCA Reagent B, 25 ml; Compatibility Reagent, 10×20 mg; Reconstitution Buffer, 15 ml; Albumin Standard Ampoules, 2 mg/ml, 10×1 ml. Microplate BCA Protein Assay Kit - RAC contains sufficient reagents for 980 microplate assays with 9 µl samples; BCA Reagent A, 250 ml; BCA Reagent B, 25 ml; Compatibility Reagent, 48×10 mg; Reconstitution Buffer, 15 ml; Albumin Standard Ampoules, 2 mg/ml, 10×1 ml; 96-well microplates, 20 plates.
Specifikace
Specifikace
- Bal.:1 KIT
- Popis:BCA protein assay kit - reducing agent compatible
- Sekvence:01
Specifikace
Accessories
Informace o produktu
- Katalogové čísloBal.DostupnostPrice
- 96-well microplate, PS, round-bottom, 5 plates per pack96-well microplate, PS, round-bottom, 5 plates per packPIER150451 SETPřihlásit se pro dostupnost1 * 1 SET - 1 960,00 Kč



