Orders LinkContactUsLinkComponent
Biopsy processing/embedding cassettes, Micromesh™ M507
Lososová
  720-0976
Biopsy processing/embedding cassettes, Micromesh™ M507
  720-0976
Biopsy processing/embedding cassettes, Micromesh™ M507
Biopsy processing/embedding cassettes, Micromesh™ M507
Lososová
10 610,00 Kč
1 * 1.000 KS
Acetal polymer. Biopsy processing/embedding cassettes, Micromesh™ M507.
  • No need for biopsy pads
  • Large square compartment, 27 mm², suitable for needle biopsies
  • 1676 square openings (0,38 mm²) allow improved fluid exchange
  • Integral lid
  • Anterior writing area 45° angle suitable for some labelling systems
  • Barva:
    Lososová