Orders LinkContactUsLinkComponent
Embedding rings
Embedding rings
Bílá
Katalogové číslo 720-0815
Dodavatel:   Simport Scientific
Embedding rings
Katalogové číslo 720-0815
Dodavatel:  Simport Scientific
Embedding rings
Embedding rings
Bílá
Přejít na:
Price
3 080,00 Kč
Přihlásit se pro dostupnost
1 * 1.000 KS
Product Details & Documents
PS. These embedding rings are suitable for holding and identifying tissue sample blocks and fit well in microtome chuck adapters.
  • The etched writing surface on the ring is marked with an identification number and placed on top of the sample block
  • Additional paraffin is poured into the base mould to cement the ring onto the tissue block
  • The embedding ring securely holds the tissue sample in the microtome chuck adapter for sectioning and then identifies the sample while in storage
Specifikace
  • Barva:
    Bílá